This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187310_m1
See other PHACTR4 GE Assays ›
Gene Symbol:
Phactr4
Gene Name:
phosphatase and actin regulator 4
Gene Aliases:
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
Chromosome Location:
Chr.4: 132355922 - 132422446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187310_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phactr4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175306.4 NP_780515.2 4 - 5 415 95
GenBank mRNA AK049209.1 - 4 - 5 378 95
BC075672.1 - 4 - 5 377 30549496 95
BC096033.1 - 4 - 5 334 6854767 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003670.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phactr4

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
neural tube closure
Rho protein signal transduction
multicellular organism development
nervous system development
actin cytoskeleton organization
positive regulation of catalytic activity
enteric nervous system development
regulation of cell cycle
closure of optic fissure
negative regulation of integrin-mediated signaling pathway
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity
protein phosphatase type 1 activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030105.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030105.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030105.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030105.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030105.mm.1 TC0400003670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030104.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030104.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030104.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030104.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030104.mm.1 TC0400003670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030107.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030107.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030107.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030107.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030107.mm.1 TC0400003670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030106.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030106.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030106.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030106.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030106.mm.1 TC0400003670.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030103.mm.1 NM_001161797.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030103.mm.1 NM_001161797.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030103.mm.1 NM_175306.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030103.mm.1 NM_175306.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030103.mm.1 TC0400003670.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0