This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01186577_m1
See other 0610040F04RIK GE Assays ›
Gene Symbol:
0610040F04Rik
Gene Name:
RIKEN cDNA 0610040F04 gene
Gene Aliases:
-
Chromosome Location:
Chr.6: 108577036 - 108660934 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01186577_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 0610040F04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040757.1 3 - 4 254 69
GenBank mRNA AK002866.1 - 3 - 4 255 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY703364.1 - 3 - 4 255 Chr. 6 - 108610333 - 108623167 [-] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream inositol 1,4,5-trisphosphate receptor 1 Itpr1 D6Pas2,
ENSMUSG00000072853,
Gm10429,
IP3R1,
InsP3R,
Ip3r,
Itpr-1,
P400,
Pcp-1,
Pcp1,
opt
-61.2kb Anti-sense Chr. 6 - 108213083 - 108551116 on NCBI Build GRCm38
Downstream RIKEN cDNA 4833447P13 gene 4833447P13Rik 44.7kb Sense Chr. 6 - 108657906 - 108659913 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002886.mm.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600022169.mm.1 NR_040757.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022169.mm.1 NR_040757.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022169.mm.1 TC0600002886.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022170.mm.1 NR_040757.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022170.mm.1 NR_040757.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022170.mm.1 TC0600002886.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022168.mm.1 NR_040757.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022168.mm.1 NR_040757.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022168.mm.1 TC0600002886.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 711322 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0