This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185272_g1
See other AIM1L GE Assays ›
Gene Symbol:
Aim1l
Gene Name:
absent in melanoma 1-like
Gene Aliases:
-
Chromosome Location:
Chr.4: 134060728 - 134092504 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185272_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aim1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162970.1 NP_001156442.1 9 - 10 3322 86
GenBank mRNA AK145690.1 - 9 - 10 3328 86
BC042709.1 - 4 - 5 343 86
BC150738.1 - 8 - 9 3251 9087649 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001626.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400012849.mm.1 NM_001162970.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400012849.mm.1 NM_001162970.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400012849.mm.1 TC0400001626.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0