This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184450_m1
See other FKBP4 GE Assays ›
Gene Symbol:
Fkbp4
Gene Name:
FK506 binding protein 4
Gene Aliases:
59kDa, AL022792, AW208983, FKBP-4, FKBP-52, FKBP52, FKPB52, p59
Chromosome Location:
Chr.6: 128430107 - 128438631 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184450_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fkbp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010219.3 NP_034349.1 3 - 4 512 99
GenBank mRNA AK051433.1 - 538 99
AK083912.1 - 3 - 4 512 99
AK089972.1 - 3 - 4 521 99
AK160202.1 - 3 - 4 461 99
BC003447.1 - 3 - 4 529 2651490 99
X17069.2 - 3 - 4 378 99
X70887.1 - 3 - 4 409 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003205.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fkbp4

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
steroid hormone receptor complex assembly
copper ion transport
embryo implantation
androgen receptor signaling pathway
prostate gland development
negative regulation of microtubule polymerization
protein complex localization
male sex differentiation
reproductive structure development
chaperone-mediated protein folding
peptidyl-prolyl cis-trans isomerase activity
protein binding
ATP binding
GTP binding
FK506 binding
isomerase activity
heat shock protein binding
copper-dependent protein binding
glucocorticoid receptor binding
poly(A) RNA binding
tau protein binding
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600025005.mm.1 NM_010219.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600025005.mm.1 NM_010219.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600025005.mm.1 TC0600003205.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0