This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184312_m1
See other SLC6A13 GE Assays ›
Gene Symbol:
Slc6a13
Gene Name:
solute carrier family 6 (neurotransmitter transporter, GABA), member 13
Gene Aliases:
GAT3, Gabt3, Gat2
Chromosome Location:
Chr.6: 121300296 - 121337724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184312_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144512.2 NP_653095.1 2 - 3 244 87
GenBank mRNA AK078399.1 - 2 - 3 251 87
AK149557.1 - 2 - 3 252 87
BC023117.1 - 2 - 3 232 4240641 87
BC029637.1 - 2 - 3 244 4195373 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001370.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a13

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
neurotransmitter transport
gamma-aminobutyric acid transport
sodium ion transmembrane transport
neurotransmitter:sodium symporter activity
gamma-aminobutyric acid:sodium symporter activity
symporter activity
neurotransmitter binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600010905.mm.1 NM_144512.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010905.mm.1 NM_144512.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010905.mm.1 TC0600001370.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0