This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184311_m1
See other SLC6A13 GE Assays ›
Gene Symbol:
Slc6a13
Gene Name:
solute carrier family 6 (neurotransmitter transporter, GABA), member 13
Gene Aliases:
GAT3, Gabt3, Gat2
Chromosome Location:
Chr.6: 121300296 - 121337724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184311_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc6a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144512.2 NP_653095.1 13 - 14 1553 97
GenBank mRNA AK149557.1 - 13 - 14 1561 97
BC021606.1 - 4 - 5 411 97
BC023117.1 - 13 - 14 1541 4240641 97
BC029637.1 - 13 - 14 1553 4195373 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001370.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc6a13

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
neurotransmitter transport
gamma-aminobutyric acid transport
sodium ion transmembrane transport
neurotransmitter:sodium symporter activity
gamma-aminobutyric acid:sodium symporter activity
symporter activity
neurotransmitter binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600010918.mm.1 NM_144512.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010918.mm.1 NM_144512.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010918.mm.1 TC0600001370.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0