This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183622_m1
See other TGFA GE Assays ›
Gene Symbol:
Tgfa
Gene Name:
transforming growth factor alpha
Gene Aliases:
wa-1, wa1
Chromosome Location:
Chr.6: 86195251 - 86275449 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183622_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031199.3 NP_112476.1 3 - 4 473 91
GenBank mRNA AK010785.1 - 3 - 4 501 91
AK162680.1 - 3 - 4 238 91
AK166321.1 - 3 - 4 406 91
BC132185.1 - 3 - 4 271 40130462 91
BC132211.1 - 3 - 4 271 40130488 91
M92420.1 - 4 - 5 223 91
U65016.1 - 3 - 4 308 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tgfa

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
angiogenesis
epidermal growth factor receptor signaling pathway
positive regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of mitotic nuclear division
negative regulation of cellular process
positive regulation of epithelial cell proliferation
positive regulation of cell division
mammary gland alveolus development
hepatocyte proliferation
glycoprotein binding
epidermal growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0