This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01182427_m1
See other PRICKLE2 GE Assays ›
Gene Symbol:
Prickle2
Gene Name:
prickle homolog 2 (Drosophila)
Gene Aliases:
6230400G14Rik, 6720451F06Rik, mpk2
Chromosome Location:
Chr.6: 92370891 - 92706335 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01182427_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prickle2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081146.2 NP_001074615.1 3 - 4 355 89
NM_001134459.1 NP_001127931.1 2 - 3 434 89
NM_001134460.2 NP_001127932.1 2 - 3 756 89
NM_001134461.1 NP_001127933.1 2 - 3 251 89
GenBank mRNA AI850364.1 - 3 - 4 297 89
AK032631.1 - 2 - 3 759 89
AK147576.1 - 2 - 3 435 89
AK147628.1 - 2 - 3 435 89
BB857589.1 - 3 - 4 357 89
BC050793.1 - 2 - 3 251 89
BC099897.1 - 3 - 4 326 30621797 89
BC145754.1 - 2 - 3 273 40131057 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002745.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

neuron projection development
establishment or maintenance of epithelial cell apical/basal polarity
protein binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600021380.mm.1 NM_001134460.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021380.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021384.mm.1 NM_001081146.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021384.mm.1 NM_001081146.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021384.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021381.mm.1 NM_001134459.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021381.mm.1 NM_001134459.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021381.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021383.mm.1 NM_001081146.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021383.mm.1 NM_001081146.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021383.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021379.mm.1 NM_001134460.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021379.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021386.mm.1 NM_001081146.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021386.mm.1 NM_001081146.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021386.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001081146.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001081146.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001134459.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001134459.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001134460.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001134461.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 NM_001134461.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021376.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001081146.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001081146.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001134459.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001134459.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001134460.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001134461.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 NM_001134461.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021377.mm.1 TC0600002745.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600021382.mm.1 NM_001134461.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021382.mm.1 NM_001134461.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021382.mm.1 TC0600002745.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 438218 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0