This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181330_g1
See other ZMIZ2 GE Assays ›
Gene Symbol:
Zmiz2
Gene Name:
zinc finger, MIZ-type containing 2
Gene Aliases:
Zimp7
Chromosome Location:
Chr.11: 6389064 - 6406162 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181330_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zmiz2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005867.1 NP_001005867.1 12 - 13 1826 60
NM_028601.3 NP_082877.2 12 - 13 1922 60
GenBank mRNA BC024081.1 - 11 - 12 1672 60
BC034541.1 - 2 - 3 268 60
BC040745.1 - 3 - 4 381 60
BC060652.1 - 12 - 13 1764 30354393 60
BC063069.1 - 4 - 5 527 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000094.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zmiz2

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of transcription from RNA polymerase II promoter
zinc ion binding
ligand-dependent nuclear receptor transcription coactivator activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100001019.mm.1 NM_001005867.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001019.mm.1 NM_001005867.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001019.mm.1 NM_028601.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100001019.mm.1 TC1100000094.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0