This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01180264_m1
See other MTIF2 GE Assays ›
Gene Symbol:
Mtif2
Gene Name:
mitochondrial translational initiation factor 2
Gene Aliases:
2310038D14Rik, 2410112O06Rik, IF-2(Mt), IF-2mt, IF2(mt)
Chromosome Location:
Chr.11: 29526379 - 29545452 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01180264_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtif2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282118.1 NP_001269047.1 12 - 13 2050 95
NM_001282119.1 NP_001269048.1 12 - 13 1972 95
NM_001282120.1 NP_001269049.1 12 - 13 2414 95
NM_133767.3 NP_598528.2 12 - 13 2328 95
GenBank mRNA AK136344.1 - 12 - 13 2012 95
AK146961.1 - 12 - 13 2268 95
BC066819.1 - 12 - 13 2308 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000296.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mtif2

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
biological_process
nucleotide binding
translation initiation factor activity
GTPase activity
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100002518.mm.1 NM_001282118.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002518.mm.1 NM_001282119.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002518.mm.1 NM_001282120.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002518.mm.1 NM_133767.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002518.mm.1 TC1100000296.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0