This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179675_m1
See other PKIB GE Assays ›
Gene Symbol:
Pkib
Gene Name:
protein kinase inhibitor beta, cAMP dependent, testis specific
Gene Aliases:
Pkibeta, Prkacn2
Chromosome Location:
Chr.10: 57631981 - 57741112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179675_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pkib
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039050.1 NP_001034139.1 2 - 3 517 156
NM_001039051.2 NP_001034140.1 2 - 3 323 156
NM_001039052.1 NP_001034141.1 2 - 3 183 156
NM_001039053.2 NP_001034142.2 3 - 4 219 156
NM_001190402.1 NP_001177331.1 2 - 3 421 156
NM_008863.4 NP_032889.3 2 - 3 323 264
GenBank mRNA AK005281.1 - 2 - 3 255 156
AK083875.1 - 2 - 3 224 264
AK156950.1 - 2 - 3 210 156
AK157521.1 - 2 - 3 211 156
BC052351.1 - 2 - 3 517 156
BC061162.1 - 2 - 3 183 156
BC087894.1 - 2 - 3 408 6771715 156
L02241.1 - 2 - 3 328 262

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK083875.1, AK156950.1, AK157521.1, BC061162.1, BC087894.1, L02241.1, NM_001039050.1, NM_001039051.2, NM_001039052.1, NM_001039053.2, NM_001190402.1, NM_008863.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pkib

Panther Classification:

Molecular Function -

kinase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
negative regulation of cAMP-dependent protein kinase activity
protein kinase inhibitor activity
cAMP-dependent protein kinase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003289.mm.1 NM_001039050.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003289.mm.1 NM_001039050.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003289.mm.1 NM_001190402.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003289.mm.1 NM_001190402.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003289.mm.1 TC1000000506.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003290.mm.1 NM_001039050.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003290.mm.1 NM_001039050.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003290.mm.1 TC1000000506.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039050.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039050.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039051.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039051.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039052.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039052.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039053.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001039053.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001190402.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_001190402.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_008863.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 NM_008863.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000003293.mm.1 TC1000000506.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000003287.mm.1 NM_001039053.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003287.mm.1 NM_001039053.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003287.mm.1 TC1000000506.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0