This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01178812_m1
See other BCKDHA GE Assays ›
Gene Symbol:
Bckdha
Gene Name:
branched chain ketoacid dehydrogenase E1, alpha polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.7: 25629852 - 25658784 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01178812_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bckdha
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007533.5 NP_031559.3 4 - 5 510 71
GenBank mRNA AK002435.1 - 4 - 5 498 71
AK154734.1 - 4 - 5 500 71
BC003787.1 - 4 - 5 522 3600452 71
L47335.1 - 4 - 5 494 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002639.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bckdha

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
branched-chain amino acid catabolic process
oxidation-reduction process
alpha-ketoacid dehydrogenase activity
3-methyl-2-oxobutanoate dehydrogenase (2-methylpropanoyl-transferring) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptor
protein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022803.mm.1 NM_007533.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022803.mm.1 NM_007533.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700022803.mm.1 TC0700002639.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0