This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177510_m1
See other SPECC1L GE Assays ›
Gene Symbol:
Specc1l
Gene Name:
sperm antigen with calponin homology and coiled-coil domains 1-like
Gene Aliases:
4930470P14Rik, 4932439K10Rik, 9530057A13Rik, AI317249, Cytsa, mKIAA0376
Chromosome Location:
Chr.10: 75212073 - 75312400 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177510_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Specc1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145826.1 NP_001139298.1 14 - 15 3247 69
NM_153406.3 NP_700455.3 11 - 12 3018 69
GenBank mRNA AB093236.1 - 14 - 15 3247 69
AY884297.1 - 11 - 12 3019 69
BC037464.1 - 8 - 9 2105 4194868 69
BC072595.1 - 10 - 11 2857 6408832 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000710.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Specc1l

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of microtubule depolymerization
cell cycle
cell adhesion
cell migration
actin cytoskeleton organization
negative regulation of actin filament depolymerization
cell division
face morphogenesis
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000005038.mm.1 NM_001145826.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005038.mm.1 NM_001145826.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005038.mm.1 NM_153406.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005038.mm.1 NM_153406.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000005038.mm.1 TC1000000710.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000005037.mm.1 NM_001145826.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005037.mm.1 NM_001145826.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005037.mm.1 NM_153406.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005037.mm.1 NM_153406.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000005037.mm.1 TC1000000710.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0