Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900002728.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002728.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002728.mm.1 |
TC0900000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002729.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002729.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002729.mm.1 |
TC0900000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002730.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002730.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002730.mm.1 |
TC0900000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002731.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002731.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002731.mm.1 |
TC0900000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002732.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002732.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002732.mm.1 |
TC0900000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002733.mm.1 |
NM_020518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002733.mm.1 |
NM_020518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002733.mm.1 |
TC0900000369.mm.1 |
MTA TC |