This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176084_m1
See other HCN4 GE Assays ›
Gene Symbol:
Hcn4
Gene Name:
hyperpolarization-activated, cyclic nucleotide-gated K+ 4
Gene Aliases:
Bcng3, Hcn3
Chromosome Location:
Chr.9: 58823512 - 58860955 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176084_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hcn4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081192.1 NP_001074661.1 1 - 2 779 79
GenBank mRNA AF064874.1 - 1 - 2 246 79
BC158106.1 - 1 - 2 879 9088182 79
BC158108.1 - 1 - 2 879 9088183 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000742.mm.1

Set Membership:

Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
regulation of heart rate
regulation of membrane depolarization
transport
ion transport
cation transport
potassium ion transport
sodium ion transport
regulation of heart contraction
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
transmembrane transport
regulation of cardiac muscle contraction
cellular response to cAMP
cellular response to cGMP
potassium ion transmembrane transport
SA node cell action potential
regulation of heart rate by cardiac conduction
cation transmembrane transport
nucleotide binding
ion channel activity
intracellular cAMP activated cation channel activity
voltage-gated ion channel activity
voltage-gated sodium channel activity
voltage-gated potassium channel activity
potassium channel activity
sodium channel activity
protein binding
cAMP binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900005469.mm.1 TC0900000742.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0