This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174910_m1
See other TRIP4 GE Assays ›
Gene Symbol:
Trip4
Gene Name:
thyroid hormone receptor interactor 4
Gene Aliases:
4930558E03Rik, ASC-1, Asc1, BB191711
Chromosome Location:
Chr.9: 65828926 - 65908809 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174910_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trip4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019797.4 NP_062771.2 14 - 15 2535 90
GenBank mRNA AF197574.1 - 11 - 12 1594 90
AF539614.1 - 14 - 15 2418 90
AK146298.1 - 11 - 12 1602 90
BC021316.1 - 11 - 12 1607 5027899 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002519.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trip4

Panther Classification:

Molecular Function -

transcription cofactor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
intracellular estrogen receptor signaling pathway
positive regulation of transcription, DNA-templated
toxin transport
protease binding
transcription coactivator activity
zinc ion binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
histone acetyltransferase binding
ubiquitin-like protein ligase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900021093.mm.1 NM_019797.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900021093.mm.1 NM_019797.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900021093.mm.1 TC0900002519.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900021094.mm.1 NM_019797.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900021094.mm.1 NM_019797.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900021094.mm.1 TC0900002519.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0