This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172791_mH
See other GNA11 GE Assays ›
Gene Symbol:
Gna11
Gene Name:
guanine nucleotide binding protein, alpha 11
Gene Aliases:
Dsk7, E430025L19
Chromosome Location:
Chr.10: 81528732 - 81545046 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172791_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gna11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010301.3 NP_034431.1 1 - 2 257 88
GenBank mRNA AK143685.1 - 1 - 2 258 88
BC011169.1 - 1 - 2 256 4206878 88
M55411.1 - 1 - 2 194 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002547.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gna11

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
action potential
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
glutamate receptor signaling pathway
heart development
regulation of melanocyte differentiation
developmental pigmentation
phospholipase C-activating dopamine receptor signaling pathway
cellular response to pH
nucleotide binding
G-protein coupled receptor binding
GTPase activity
signal transducer activity
GTP binding
guanyl nucleotide binding
G-protein beta/gamma-subunit complex binding
type 2A serotonin receptor binding
protein complex binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000019211.mm.1 NM_010301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000019211.mm.1 NM_010301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000019211.mm.1 TC1000002547.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000019210.mm.1 NM_010301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000019210.mm.1 NM_010301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000019210.mm.1 TC1000002547.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0