This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171418_m1
See other PDE4A GE Assays ›
Gene Symbol:
Pde4a
Gene Name:
phosphodiesterase 4A, cAMP specific
Gene Aliases:
D9Ertd60e, Dpde2
Chromosome Location:
Chr.9: 21165683 - 21213248 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171418_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183408.3 NP_899668.1 1 - 2 516 78
GenBank mRNA BC139405.1 - 1 - 2 356 8862427 78
BC139408.1 - 1 - 2 356 8862430 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000189.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde4a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
sensory perception of smell
regulation of protein kinase A signaling
cellular response to drug
regulation of cAMP-mediated signaling
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
cAMP binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001244.mm.1 NM_183408.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001244.mm.1 NM_183408.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900001244.mm.1 TC0900000189.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001249.mm.1 NM_183408.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001249.mm.1 NM_183408.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900001249.mm.1 TC0900000189.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900001245.mm.1 NM_183408.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001245.mm.1 NM_183408.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900001245.mm.1 TC0900000189.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 649183 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0