This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171335_g1
See other ARHGAP33 GE Assays ›
Gene Symbol:
Arhgap33
Gene Name:
Rho GTPase activating protein 33
Gene Aliases:
AW324378, NOMA-GAP, Snx26, TCGAP
Chromosome Location:
Chr.7: 30522226 - 30535092 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171335_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289670.1 NP_001276599.1 8 - 9 835 94
NM_001289682.1 NP_001276611.1 9 - 10 918 94
NM_178252.3 NP_839983.1 9 - 10 945 94
GenBank mRNA AK142180.1 - 8 - 9 836 94
AY217764.1 - 8 - 9 778 94
BC061471.1 - 2 - 3 122 5698780 94
BC065086.1 - 9 - 10 894 6850475 94
BC065166.1 - 9 - 10 892 6836510 94
BC066047.1 - 9 - 10 870 6822191 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC061471.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap33

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

transport
signal transduction
protein transport
positive regulation of GTPase activity
GTPase activator activity
protein binding
phosphatidylinositol binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700024552.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024552.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024552.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024552.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024555.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024555.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024556.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024556.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024556.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024556.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024545.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024545.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024545.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024545.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024548.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024548.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024548.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024548.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024550.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024550.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024550.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024550.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024546.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024546.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024546.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024546.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024547.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024547.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024547.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024547.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024551.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024551.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024551.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024551.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024544.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024544.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024544.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024544.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024549.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024549.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024549.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024549.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024559.mm.1 NM_001289670.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024559.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024559.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024559.mm.1 TC0700002763.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024553.mm.1 NM_001289682.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024553.mm.1 NM_178252.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700024553.mm.1 TC0700002763.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 729575 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0