This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170712_m1
See other MAP2K5 GE Assays ›
Gene Symbol:
Map2k5
Gene Name:
mitogen-activated protein kinase kinase 5
Gene Aliases:
AI324775, AI428457, MEK5, Mapkk5, Prkmk5
Chromosome Location:
Chr.9: 63163768 - 63378438 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170712_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Map2k5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011840.2 NP_035970.1 17 - 18 1667 77
GenBank mRNA AB019374.1 - 17 - 18 1668 77
AK013856.1 - 9 - 10 510 77
AK159261.1 - 17 - 18 1717 77
AK178825.1 - 2 - 3 59 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002472.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Map2k5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPK activity
protein phosphorylation
heart development
phosphorylation
positive regulation of cell growth
negative regulation of NF-kappaB transcription factor activity
negative regulation of heterotypic cell-cell adhesion
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
positive regulation of protein metabolic process
negative regulation of response to cytokine stimulus
ERK5 cascade
cellular response to growth factor stimulus
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of chemokine (C-X-C motif) ligand 2 production
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900020698.mm.1 NM_011840.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900020698.mm.1 NM_011840.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900020698.mm.1 TC0900002472.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0