This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170652_m1
See other PAK2 GE Assays ›
Gene Symbol:
Pak2
Gene Name:
p21 protein (Cdc42/Rac)-activated kinase 2
Gene Aliases:
5330420P17Rik, A130002K10Rik, AI836325, D16Ertd269e, PAK-2, mKIAA4182
Chromosome Location:
Chr.16: 32016290 - 32079342 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170652_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pak2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177326.3 NP_796300.1 6 - 7 856 71
GenBank mRNA AK037275.1 - 6 - 7 857 71
AY167030.1 - 5 - 6 582 71
BC063068.1 - 844 71
BC086650.1 - 6 - 7 787 6849008 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001497.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pak2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
Rho protein signal transduction
regulation of mitotic cell cycle
metabolic process
phosphorylation
cell migration
peptidyl-serine phosphorylation
signal transduction by protein phosphorylation
actin cytoskeleton organization
stress-activated protein kinase signaling cascade
activation of protein kinase activity
regulation of growth
negative regulation of apoptotic process
regulation of MAPK cascade
protein autophosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
dendritic spine development
positive regulation of protein tyrosine kinase activity
cellular response to organic cyclic compound
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
nucleotide binding
catalytic activity
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein tyrosine kinase activator activity
identical protein binding
Rac GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600012295.mm.1 NM_177326.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600012295.mm.1 TC1600001497.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0