This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170323_m1
See other ANLN GE Assays ›
Gene Symbol:
Anln
Gene Name:
anillin, actin binding protein
Gene Aliases:
1110037A17Rik, 2900037I21Rik, C78101, Scraps
Chromosome Location:
Chr.9: 22331214 - 22389696 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170323_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anln
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028390.3 NP_082666.2 12 - 13 2369 139
GenBank mRNA AK045703.1 - 9 - 10 1333 139
AK046102.1 - 1 - 2 155 139
BC030489.1 - 5 - 6 711 5373071 139
BC032164.1 - 12 - 13 2387 5346703 139

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK046102.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anln

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
hematopoietic progenitor cell differentiation
cell cycle
mitotic nuclear division
cell division
glomerular visceral epithelial cell migration
actin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016080.mm.1 NM_028390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016080.mm.1 NM_028390.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900016080.mm.1 TC0900001908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0