This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169213_m1
See other ZFP65 GE Assays ›
Gene Symbol:
Zfp65
Gene Name:
zinc finger protein 65
Gene Aliases:
KRAB5, Zfp71-rs1, pz114
Chromosome Location:
Chr.13: 67705306 - 67729173 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169213_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145622.2 NP_663597.1 1 - 2 116 68
GenBank mRNA AK041786.1 - 1 - 2 63 68
BB621429.1 - 1 - 2 117 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK041786.1, NM_145622.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp65

Panther Classification:

Molecular Function -

DNA-binding transcription factor C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300014763.mm.1 NM_145622.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014763.mm.1 NM_145622.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300014763.mm.1 TC1300002259.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014764.mm.1 NM_145622.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014764.mm.1 NM_145622.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300014764.mm.1 TC1300002259.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300014758.mm.1 NM_145622.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300014758.mm.1 NM_145622.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300014758.mm.1 TC1300002259.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0