This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168324_m1
See other CALU GE Assays ›
Gene Symbol:
Calu
Gene Name:
calumenin
Gene Aliases:
9530075H20Rik
Chromosome Location:
Chr.6: 29348106 - 29376675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168324_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Calu
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285412.1 NP_001272341.1 2 - 3 149 72
NM_007594.4 NP_031620.1 2 - 3 332 72
GenBank mRNA AK135295.1 - 2 - 3 257 72
AK148178.1 - 2 - 3 285 72
AK160795.1 - 2 - 3 285 72
AK161807.1 - 2 - 3 289 72
AK163674.1 - 2 - 3 320 72
BC051423.1 - 2 - 3 293 3989906 72
U81829.1 - 2 - 3 258 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001285412.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Calu

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
enzyme inhibitor activity
calcium ion binding
enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600001472.mm.1 NM_001285412.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001472.mm.1 NM_007594.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600001472.mm.1 TC0600000229.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600001473.mm.1 NM_001285412.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600001473.mm.1 NM_007594.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600001473.mm.1 TC0600000229.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 349954 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0