This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168049_m1
See other PRL2A1 GE Assays ›
Gene Symbol:
Prl2a1
Gene Name:
prolactin family 2, subfamily a, member 1
Gene Aliases:
PLP-M, Prlpm
Chromosome Location:
Chr.13: 27801655 - 27808716 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168049_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prl2a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019991.1 NP_064375.1 2 - 3 254 92
GenBank mRNA AB032428.1 - 2 - 3 275 92
AF226610.1 - 2 - 3 210 92
AF234636.1 - 2 - 3 254 92
AF272368.1 - 2 - 3 206 92
AK005424.1 - 2 - 3 231 92
BC099458.1 - 2 - 3 250 30790018 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000304.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prl2a1

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
hormone activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300001889.mm.1 NM_019991.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001889.mm.1 NM_019991.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300001889.mm.1 TC1300000304.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0