Assay Details
Gene Symbol: Cct8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198621.2 | NP_941023.1 | 1 | 1779 | 67 | |
GenBank mRNA | AK133375.1 | - | 1 | 1779 | 67 | |
BC050797.1 | - | 1 | 1696 | 6743432 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | AK133375.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | BC050797.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | AK133375.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | BC050797.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646933 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154639 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | AK133375.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | BC050797.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | AK133375.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | BC050797.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547837 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547842 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547835 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23547862 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154639 | - | - |
Whole Mouse Genome G4122A | A_52_P407191 | AK133375.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P407191 | BC050797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P407191 | AK133375.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P407191 | BC050797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P407191 | NM_198621.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112535 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154639 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04365625 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04365641 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04365645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23547855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04365640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04347411 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547843 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04901769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04994943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04365628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04365641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04365632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04901771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04187297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04994951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547833 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04658892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04653226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547835 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547905 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04994942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547853 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547910 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04221908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547842 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04901778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04187296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23547845 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10520302 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10520303 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1437962_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10520301 | - | - |
Mouse Genome 430 2.0 Array | 1437962_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1437962_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242891_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 242891_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500001824.mm.1 | NM_198621.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500001824.mm.1 | NM_198621.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500001824.mm.1 | TC0500000234.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500001825.mm.1 | NM_198621.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500001825.mm.1 | NM_198621.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500001825.mm.1 | TC0500000234.mm.1 | MTA TC |
Mouse Expression Set 430 | 1437962_at | - | - |
Mouse Expression Set 430 | 1437962_PM_at | - | - |
Murine Genome U74v2 Set | 166249_PM_r_at | - | - |
Murine Genome U74v2 Set | 166249_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437962_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1437962_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 855062 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2260142 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2260142 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002260142 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598594 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2598594 | - | - |
mouse-6 v1.1 (Illumina) | scl0242891.1_147-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2260142 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0242891.1_147-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260142 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2598594 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2598594 | - | - |
