This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165701_g1
See other RRM2B GE Assays ›
Gene Symbol:
Rrm2b
Gene Name:
ribonucleotide reductase M2 B (TP53 inducible)
Gene Aliases:
p53R2
Chromosome Location:
Chr.15: 37923952 - 37961108 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165701_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rrm2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199476.1 NP_955770.1 2 - 3 366 62
GenBank mRNA AK138731.1 - 2 - 3 359 62
AK164359.1 - 1 - 2 1537 62
BC058103.1 - 2 - 3 366 6827753 62

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rrm2b

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

kidney development
renal system process
DNA replication
mitochondrial DNA replication
DNA repair
cellular response to DNA damage stimulus
response to oxidative stress
deoxyribonucleoside diphosphate metabolic process
deoxyribonucleoside triphosphate metabolic process
deoxyribonucleotide biosynthetic process
oxidation-reduction process
negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
ribonucleoside-diphosphate reductase activity, thioredoxin disulfide as acceptor
oxidoreductase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500010839.mm.1 NM_199476.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500010839.mm.1 NM_199476.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500010839.mm.1 TC1500001401.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0