This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165077_m1
See other CNOT4 GE Assays ›
Gene Symbol:
Cnot4
Gene Name:
CCR4-NOT transcription complex, subunit 4
Gene Aliases:
Not4, Not4h, Not4hp
Chromosome Location:
Chr.6: 35022065 - 35133737 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165077_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnot4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164411.1 NP_001157883.1 8 - 9 1195 88
NM_001164412.1 NP_001157884.1 8 - 9 1195 88
NM_001164413.1 NP_001157885.1 8 - 9 1195 88
NM_001164414.1 NP_001157886.1 8 - 9 1204 88
NM_016877.4 NP_058573.3 8 - 9 1204 88
GenBank mRNA AK028190.1 - 8 - 9 1065 88
AK032248.1 - 8 - 9 1184 88
AK144760.1 - 8 - 9 1162 88
AK165997.1 - 8 - 9 1181 88
BC048243.1 - 8 - 9 1083 6493673 88
BC058778.1 - 8 - 9 1147 5397231 88
U71269.1 - 8 - 9 1190 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002059.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnot4

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
protein autoubiquitination
nucleotide binding
nucleic acid binding
RNA binding
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
poly(A) RNA binding
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164411.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164411.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164412.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164412.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164413.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164413.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164414.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_001164414.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_016877.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 NM_016877.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600016237.mm.1 TC0600002059.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0