This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164966_m1
See other CPQ GE Assays ›
Gene Symbol:
Cpq
Gene Name:
carboxypeptidase Q
Gene Aliases:
1190003P12Rik, 2610034C17Rik, Hls2, Lal-1, Pgcp
Chromosome Location:
Chr.15: 33083129 - 33594552 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164966_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpq
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176073.4 NP_788262.1 1 - 2 191 84
GenBank mRNA AF009513.1 - 1 - 2 72 84
AF107835.1 - 1 - 2 159 84
AK054422.1 - 1 - 2 65 84
AK075686.1 - 1 - 2 122 84
AK135096.1 - 1 - 2 178 84
AK137164.1 - 1 - 2 65 84
AK154361.1 - 1 - 2 65 84
BC010977.1 - 1 - 2 177 84
CK624628.1 - 1 - 2 191 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF009513.1, AF107835.1, AK075686.1, AK135096.1, AK137164.1, AK154361.1, NM_176073.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpq

Panther Classification:

Molecular Function -

metalloprotease protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
thyroid hormone generation
peptide catabolic process
carboxypeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
protein homodimerization activity
metal ion binding
metallodipeptidase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001649.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001649.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001649.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001649.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001649.mm.1 TC1500000222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001640.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001640.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001640.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001640.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001640.mm.1 TC1500000222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001641.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001641.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001641.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001641.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001641.mm.1 TC1500000222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001643.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001643.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001643.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001643.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001643.mm.1 TC1500000222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001642.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001642.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001642.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001642.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001642.mm.1 TC1500000222.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001648.mm.1 NM_018755.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001648.mm.1 NM_018755.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001648.mm.1 NM_176073.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001648.mm.1 NM_176073.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500001648.mm.1 TC1500000222.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 796510 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0