This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164806_m1
See other 4930557F08RIK GE Assays ›
Gene Symbol:
4930557F08Rik
Gene Name:
RIKEN cDNA 4930557F08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164806_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930557F08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016160.1 - 3 - 4 315 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715764.1 - 3 - 4 311 Chr. 15 - 15779996 - 15823426 [+] on Build GRCm38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cadherin 6 Cdh6 K-cadherin,
cad6,
cadherin-6
-2.6mb Anti-sense Chr. 15 - 13028699 - 13173689 on NCBI Build GRCm38
Downstream cadherin 9 Cdh9 905.1kb Sense Chr. 15 - 16728549 - 16857097 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838124 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139536 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46253 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728269 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630995 AK077094.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630995 AK139540.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630995 AK163350.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630995 BC044740.2 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630995 NM_001033226.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606300 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158247 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866778 AK015139.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866778 AK076690.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742096 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858613 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21686 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134269 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103276 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08294037 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08294007 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08293991 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108395 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08294037 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037867 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037873 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038043 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038046 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037875 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037877 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038055 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038050 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037872 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038049 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037880 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037886 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037871 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038042 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038048 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038051 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038035 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038036 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037884 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038053 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038059 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038054 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038056 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038057 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12450978 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037869 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038041 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037887 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038039 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037888 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30037882 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30038038 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0