This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01163943_m1
See other EGFL8 GE Assays ›
Gene Symbol:
Egfl8
Gene Name:
EGF-like domain 8
Gene Aliases:
Ng3
Chromosome Location:
Chr.17: 34613200 - 34616572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01163943_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egfl8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152922.3 NP_690886.3 1 - 2 64 90
GenBank mRNA AK148723.1 - 1 - 2 65 90
BC139484.1 - 1 - 2 33 8862506 90
BC141312.1 - 1 - 2 33 9055924 90
BC145632.1 - 1 - 2 33 9054200 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK148723.1,BC139484.1,BC141312.1,BC145632.1,NM_152922.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Egfl8

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700017928.mm.1 NM_152922.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700017928.mm.1 NM_152922.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700017928.mm.1 TC1700002812.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700017920.mm.1 NM_152922.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700017920.mm.1 NM_152922.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700017920.mm.1 TC1700002812.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 763034 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0