This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01163699_m1
See other SEC23IP GE Assays ›
Gene Symbol:
Sec23ip
Gene Name:
Sec23 interacting protein
Gene Aliases:
D7Ertd373e, p125
Chromosome Location:
Chr.7: 128744862 - 128784836 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01163699_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec23ip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029982.2 NP_001025153.2 3 - 4 1051 81
GenBank mRNA AK048053.1 - 3 - 4 980 81
BC098506.1 - 3 - 4 940 30014037 81
BC116304.1 - 2 - 3 432 81
BC116305.1 - 2 - 3 432 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001893.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec23ip

Panther Classification:

Molecular Function -

hydrolase lipase phospholipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
spermatid development
single fertilization
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700016508.mm.1 NM_001029982.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016508.mm.1 NM_001029982.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700016508.mm.1 TC0700001893.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0