This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01163465_m1
See other TMEM80 GE Assays ›
Gene Symbol:
Tmem80
Gene Name:
transmembrane protein 80
Gene Aliases:
5530601I19Rik
Chromosome Location:
Chr.7: 141327743 - 141337156 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01163465_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001141950.1 NP_001135422.1 3 - 4 297 88
GenBank mRNA AK134821.1 - 3 - 4 298 88
AK169083.1 - 3 - 4 256 88
BC022756.1 - 3 - 4 241 4922223 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK134821.1, AK169083.1, BC022756.1, NM_001141950.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0