This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01163374_m1
See other SH2B1 GE Assays ›
Gene Symbol:
Sh2b1
Gene Name:
SH2B adaptor protein 1
Gene Aliases:
AI425885, C530001K22Rik, Irip, Psm, SH2-B, SH2-Bb, Sh2bpsm1, mKIAA1299
Chromosome Location:
Chr.7: 126466993 - 126475684 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01163374_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh2b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289541.1 NP_001276470.1 8 - 9 2164 108
NM_001289542.1 NP_001276471.1 8 - 9 2541 108
GenBank mRNA AF380422.1 - 8 - 9 2235 108
BC051978.1 - 8 - 9 2084 6401205 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004280.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
lamellipodium assembly
intracellular signal transduction
positive regulation of mitotic nuclear division
regulation of DNA biosynthetic process
signal transducer activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700035686.mm.1 NM_001289541.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035686.mm.1 NM_001289542.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035686.mm.1 NM_011363.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035686.mm.1 TC0700004280.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0