This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01162828_m1
See other PLEKHA7 GE Assays ›
Gene Symbol:
Plekha7
Gene Name:
pleckstrin homology domain containing, family A member 7
Gene Aliases:
A430081P20Rik, AI414265
Chromosome Location:
Chr.7: 116124118 - 116308381 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01162828_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plekha7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK040271.1 - 15 - 16 2238 75
AK146866.1 - 2429 75
AK160810.1 - 15 - 16 2318 75
BC060678.1 - 14 - 15 2159 75
BC084587.1 - 15 - 16 2333 3602481 75
EU380771.1 - 2455 75
EU380772.1 - 2513 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0