This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01161255_m1
See other SERPINB9 GE Assays ›
Gene Symbol:
Serpinb9
Gene Name:
serine (or cysteine) peptidase inhibitor, clade B, member 9
Gene Aliases:
BB283241, CAP-3, CAP3, PI-9, PI9, Spi6, ovalbumin
Chromosome Location:
Chr.13: 32997061 - 33017955 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01161255_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serpinb9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009256.3 NP_033282.1 5 - 6 737 101
GenBank mRNA AK035624.1 - 5 - 6 676 101
AK087785.1 - 5 - 6 646 101
AK161581.1 - 5 - 6 738 101
BC029900.1 - 5 - 6 665 4925100 101
U96700.1 - 5 - 6 616 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serpinb9

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

T cell mediated cytotoxicity
acute inflammatory response to antigenic stimulus
apoptotic process
immune response
response to bacterium
positive regulation of gene expression
negative regulation of endopeptidase activity
negative regulation by symbiont of host apoptotic process
protection from natural killer cell mediated cytotoxicity
defense response to bacterium
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of T cell apoptotic process
protease binding
serine-type endopeptidase inhibitor activity
protein binding
cysteine-type endopeptidase inhibitor activity involved in apoptotic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0