This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01161207_m1
See other TMEVPG1 GE Assays ›
Gene Symbol:
Tmevpg1
Gene Name:
Theiler's murine encephalomyelitis virus persistence candidate gene 1
Gene Aliases:
AI447422
Chromosome Location:
Chr.10: 118502035 - 118556525 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01161207_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmevpg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104123.1 4 - 5 468 149
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI592225.1 - 4 - 5 469 Chr. 10 - 118502590 - 118556525 [-] on Build GRCm38 150
GenBankmRNA AA162222.1 - 4 - 5 468 Chr. 10 - 118502574 - 118556525 [-] on Build GRCm38 149

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream interferon gamma Ifng IFN-g,
Ifg
-56.7kb Anti-sense Chr. 10 - 118441047 - 118445892 on NCBI Build GRCm38
Downstream predicted gene,
33337
Gm33337 202.9kb Sense Chr. 10 - 118714522 - 118751241 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002992.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022405.mm.1 NR_104123.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022405.mm.1 TC1000002992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022406.mm.1 NR_104123.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022406.mm.1 TC1000002992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000022408.mm.1 NR_104123.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022408.mm.1 TC1000002992.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 843465 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101780451 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0