This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159967_g1
See other CD63 GE Assays ›
Gene Symbol:
Cd63
Gene Name:
CD63 antigen
Gene Aliases:
C75951, ME491, Tspan30
Chromosome Location:
Chr.10: 128908919 - 128912822 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159967_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd63
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042580.1 NP_001036045.1 5 - 6 574 106
NM_001282966.1 NP_001269895.1 5 - 6 618 106
NM_007653.3 NP_031679.1 5 - 6 533 106
GenBank mRNA AF159427.1 - 5 - 6 454 106
AK028220.1 - 5 - 6 471 106
AK146618.1 - 5 - 6 521 106
AK151702.1 - 5 - 6 499 106
AK151963.1 - 5 - 6 473 106
AK152072.1 - 5 - 6 495 106
AK152372.1 - 5 - 6 499 106
AK159875.1 - 5 - 6 496 106
AK163785.1 - 5 - 6 495 106
AK166543.1 - 5 - 6 494 106
AK166766.1 - 5 - 6 493 106
AK170344.1 - 5 - 6 528 106
AK182243.1 - 3 - 4 329 106
AK216882.1 - 3 - 4 329 106
BC008108.1 - 5 - 6 480 3157993 106
BC012212.1 - 5 - 6 480 3157993 106
BC083161.1 - 5 - 6 465 6530532 106
BP769648.1 - 3 - 4 386 106
CT010179.1 - 4 - 5 429 106
D16432.1 - 5 - 6 483 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd63

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of receptor internalization
transport
cell-matrix adhesion
negative regulation of epithelial cell migration
protein transport
cell migration
epithelial cell differentiation
cellular protein localization
endosome to melanosome transport
pigmentation
positive regulation of cell adhesion
positive regulation of endocytosis
pigment granule maturation
pigment cell differentiation
regulation of vascular endothelial growth factor signaling pathway
regulation of rubidium ion transport
positive regulation of integrin-mediated signaling pathway
protein binding
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0