This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159102_m1
See other ITGAD GE Assays ›
Gene Symbol:
Itgad
Gene Name:
integrin, alpha D
Gene Aliases:
Cd11d
Chromosome Location:
Chr.7: 128173933 - 128205521 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159102_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itgad
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029872.3 NP_001025043.3 1 - 2 81 90
GenBank mRNA AK156218.1 - 1 - 2 82 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK156218.1, NM_001029872.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Itgad

Panther Classification:

Molecular Function -

integrin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
integrin-mediated signaling pathway
heterotypic cell-cell adhesion
activated T cell proliferation
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700016326.mm.1 NM_001029872.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016326.mm.1 NM_001029872.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700016326.mm.1 TC0700001877.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700016327.mm.1 NM_001029872.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700016327.mm.1 NM_001029872.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700016327.mm.1 TC0700001877.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0