This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157399_m1
See other D130043K22RIK GE Assays ›
Gene Symbol:
D130043K22Rik
Gene Name:
RIKEN cDNA D130043K22 gene
Gene Aliases:
4930451E12Rik, Kiaa0319
Chromosome Location:
Chr.13: 24843930 - 24902159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157399_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130043K22Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK122246.1 - 15 - 16 2711 73
AK148668.1 - 13 - 14 2318 73
BC115723.1 - 15 - 16 2630 40086618 73
BC115940.1 - 15 - 16 2630 40086616 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000281.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300001707.mm.1 NM_001081051.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300001707.mm.1 TC1300000281.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300001706.mm.1 NM_001081051.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300001706.mm.1 TC1300000281.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0