This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01157051_m1
See other SRGAP1 GE Assays ›
Gene Symbol:
Srgap1
Gene Name:
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases:
4930572H05Rik, Arhgap13
Chromosome Location:
Chr.10: 121780991 - 122047315 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01157051_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081037.2 NP_001074506.1 14 - 15 1803 77
NM_001242411.1 NP_001229340.1 14 - 15 1734 77
GenBank mRNA AY057898.1 - 7 - 8 565 77
BC120892.1 - 14 - 15 40061454 77
BC120893.1 - 14 - 15 40061455 77
BU706317.1 - 3 - 4 175 77
CX569081.1 - 6 - 7 437 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
Rho protein signal transduction
cell migration
negative regulation of cell migration
positive regulation of GTPase activity
GTPase activator activity
protein binding
Rho GTPase binding
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0