This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01151011_m1
See other AP1M2 GE Assays ›
Gene Symbol:
Ap1m2
Gene Name:
adaptor protein complex AP-1, mu 2 subunit
Gene Aliases:
D9Ertd818e, [m]1B, mu1B
Chromosome Location:
Chr.9: 21294613 - 21312349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01151011_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ap1m2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110300.1 NP_001103770.1 1 - 2 125 71
NM_009678.2 NP_033808.2 1 - 2 125 71
GenBank mRNA AF067146.1 - 1 - 2 99 71
AK010478.1 - 1 - 2 104 71
AK082674.1 - 109 71
AK160161.1 - 1 - 2 92 71
BC003704.1 - 1 - 2 64 3488231 71
CJ078558.1 - 1 - 2 126 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ap1m2

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s)

transport
intracellular protein transport
protein transport
vesicle-mediated transport

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 510201 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0