This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01150868_m1
See other TIRAP GE Assays ›
Gene Symbol:
Tirap
Gene Name:
toll-interleukin 1 receptor (TIR) domain-containing adaptor protein
Gene Aliases:
AA407980, C130027E04Rik, Mal, Tlr4ap, Wyatt
Chromosome Location:
Chr.9: 35184391 - 35200291 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01150868_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tirap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177845.1 NP_001171316.1 2 - 3 231 67
NM_001177846.1 NP_001171317.1 2 - 3 213 67
GenBank mRNA AK036431.1 - 2 - 3 152 67
AK080135.1 - 2 - 3 232 67
BY740695.1 - 2 - 3 219 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK036431.1, AK080135.1, NM_001177845.1, NM_001177846.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

immune system process
inflammatory response
signal transduction
cell surface receptor signaling pathway
I-kappaB kinase/NF-kappaB signaling
activation of JUN kinase activity
response to bacterium
myeloid cell differentiation
positive regulation of B cell proliferation
response to lipopolysaccharide
regulation of interferon-beta production
regulation of interleukin-15 production
positive regulation of interleukin-12 production
positive regulation of interleukin-15 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
positive regulation of toll-like receptor 2 signaling pathway
positive regulation of toll-like receptor 3 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
toll-like receptor 5 signaling pathway
TIRAP-dependent toll-like receptor 4 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of growth of symbiont in host
innate immune response
regulation of innate immune response
positive regulation of innate immune response
positive regulation of interleukin-6 biosynthetic process
positive regulation of JNK cascade
defense response to Gram-positive bacterium
positive regulation of NF-kappaB transcription factor activity
positive regulation of ERK1 and ERK2 cascade
interleukin-1-mediated signaling pathway
3'-UTR-mediated mRNA stabilization
cellular response to bacterial lipopeptide
cellular response to lipoteichoic acid
positive regulation of neutrophil chemotaxis
positive regulation of protein homodimerization activity
positive regulation of chemokine (C-X-C motif) ligand 1 production
positive regulation of chemokine (C-X-C motif) ligand 2 production
protein kinase C binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
Toll-like receptor 4 binding
Toll-like receptor 2 binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177845.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177845.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177846.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177846.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177847.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_001177847.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_054096.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 NM_054096.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900016680.mm.1 TC0900002008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016689.mm.1 NM_001177845.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016689.mm.1 NM_001177845.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016689.mm.1 TC0900002008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177845.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177845.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177846.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177846.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177847.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_001177847.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_054096.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 NM_054096.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900016678.mm.1 TC0900002008.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016686.mm.1 NM_001177846.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016686.mm.1 NM_001177846.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016686.mm.1 NM_054096.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016686.mm.1 NM_054096.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900016686.mm.1 TC0900002008.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0