This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01150092_g1
See other PLK5 GE Assays ›
Gene Symbol:
Plk5
Gene Name:
polo-like kinase 5
Gene Aliases:
6330514A18Rik
Chromosome Location:
Chr.10: 80356459 - 80365489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01150092_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plk5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183152.3 NP_898975.2 5 - 6 667 140
GenBank mRNA AK031968.1 - 5 - 6 668 139
BC099679.1 - 5 - 6 636 6489647 140
BG297060.1 - 3 - 4 254 140

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK031968.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plk5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

defense response to tumor cell
protein phosphorylation
obsolete transformed cell apoptotic process
cellular response to DNA damage stimulus
cell cycle
negative regulation of cell proliferation
positive regulation of neuron projection development
cell differentiation
G2 DNA damage checkpoint
cell division
cellular response to growth factor stimulus
regulation of G1/S transition of mitotic cell cycle
nucleotide binding
protein kinase activity
ATP binding
kinase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0