This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01149792_m1
See other S100B GE Assays ›
Gene Symbol:
S100b
Gene Name:
S100 protein, beta polypeptide, neural
Gene Aliases:
AI850290, Bpb
Chromosome Location:
Chr.10: 76253836 - 76261319 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01149792_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009115.3 NP_033141.1 1 - 2 129 67
GenBank mRNA AK134625.1 - 1 - 2 56 67
AK135083.1 - 1 - 2 112 67
AK161945.1 - 1 - 2 105 67
BC061178.1 - 1 - 2 112 6535966 67
CX205737.1 - 1 - 2 129 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK134625.1, AK135083.1, AK161945.1, BC061178.1, NM_009115.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100b

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

energy reserve metabolic process
cellular calcium ion homeostasis
learning or memory
memory
positive regulation of cell proliferation
regulation of cell shape
regulation of cytokine biosynthetic process
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
positive regulation of synaptic transmission
negative regulation of skeletal muscle cell differentiation
receptor binding
calcium ion binding
zinc ion binding
identical protein binding
protein homodimerization activity
S100 protein binding
metal ion binding
calcium-dependent protein binding
RAGE receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000005266.mm.1 NM_009115.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005266.mm.1 NM_009115.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000005266.mm.1 TC1000000736.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000005267.mm.1 NM_009115.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005267.mm.1 NM_009115.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000005267.mm.1 TC1000000736.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0