Assay Details
Gene Symbol: Gstt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010361.2 | NP_034491.2 | 4 - 5 | 1121 | 70 | |
GenBank mRNA | AK002392.1 | - | 4 - 5 | 651 | 70 | |
AK158939.1 | - | 1112 | 70 | |||
BC012707.1 | - | 4 - 5 | 679 | 3994154 | 70 | |
X98056.1 | - | 4 - 5 | 526 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258887 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258886 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258890 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258888 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258889 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | U48420.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | X98056.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | U48420.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | X98056.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508137 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241869 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | U48420.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | X98056.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | AK002392.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | AK158939.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | BC012707.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | U48420.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | X98056.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | AK002392.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | AK158939.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | U48420.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | X98056.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | U48420.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | X98056.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258901 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258858 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258874 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258915 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258888 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27258879 | - | - |
Whole Mouse Genome G4122A | A_51_P350048 | U48420.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | X98056.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | U48420.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | X98056.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | AK002392.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | AK158939.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P350048 | BC012707.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128244 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08727897 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08831149 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08727906 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08727893 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08727896 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258886 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258890 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258887 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258889 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27258888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27258856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08727890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08790874 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08462693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413330 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08462690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08225819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258934 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08727891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08441763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08462695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08719680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08386042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08790857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08386043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08727896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08727901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27258953 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10370015 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370016 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370018 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370019 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370017 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10370020 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417883_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10370013 | - | - |
Mouse Genome 430 2.0 Array | 1417883_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1417883_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14872_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14872_at | - | - |
Mouse Expression Set 430 | 1417883_at | - | - |
Mouse Expression Set 430 | 1417883_PM_at | - | - |
Murine Genome U74v2 Set | 104603_PM_at | - | - |
Murine Genome U74v2 Set | 104603_at | - | - |
Mouse Genome 430A 2.0 Array | 1417883_at | - | - |
Mouse Genome 430A 2.0 Array | 1417883_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417883_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417883_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 634462 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1580519 | - | - |
MouseWG-6 v2 Expression BeadChip | 106650102 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1580519 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 106650102 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001580519 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689880 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2554549 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2689880 | - | - |
mouse-6 v1.1 (Illumina) | scl014872.2_19-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106650102 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1580519 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl014872.2_19-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106650102 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1580519 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2689880 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689880 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2554549 | - | - |
