This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01145828_m1
See other ATP8B3 GE Assays ›
Gene Symbol:
Atp8b3
Gene Name:
ATPase, class I, type 8B, member 3
Gene Aliases:
1700042F02Rik, 1700056N23Rik, ATPIK, SAPLT
Chromosome Location:
Chr.10: 80519584 - 80539150 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01145828_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp8b3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026094.3 NP_080370.2 9 - 10 759 70
GenBank mRNA AY364445.1 - 9 - 10 759 70
BC118977.1 - 9 - 10 759 40098417 70
BC118978.1 - 9 - 10 759 40098419 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002503.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp8b3

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
Golgi organization
binding of sperm to zona pellucida
phospholipid transport
phospholipid translocation
nucleotide binding
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000018590.mm.1 NM_026094.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018590.mm.1 NM_026094.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018590.mm.1 TC1000002503.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0