This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01143729_m1
See other ASCC1 GE Assays ›
Gene Symbol:
Ascc1
Gene Name:
activating signal cointegrator 1 complex subunit 1
Gene Aliases:
1810015P09Rik, AI550520, ASC1p50, CGI-18
Chromosome Location:
Chr.10: 60002493 - 60099990 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01143729_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ascc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199187.2 NP_001186116.1 9 - 10 1061 117
NM_026937.3 NP_081213.1 9 - 10 1074 117
GenBank mRNA AK007519.1 - 9 - 10 1058 117
AK139944.1 - 9 - 10 1061 117
AK171958.1 - 9 - 10 1035 117
BC030905.1 - 9 - 10 1040 4503445 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000541.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ascc1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleic acid binding
transcription coactivator activity
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003653.mm.1 NM_001199187.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003653.mm.1 NM_001199187.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003653.mm.1 NM_026937.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003653.mm.1 NM_026937.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003653.mm.1 TC1000000541.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0