This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01139030_mH
See other ANKHD1 GE Assays ›
Gene Symbol:
Ankhd1
Gene Name:
ankyrin repeat and KH domain containing 1
Gene Aliases:
1110004O12Rik, 4933432B13Rik, 9130019P20Rik, A530027J04Rik, A630021B20Rik, AA571404, Mask, mFLJ00246, mKIAA1085
Chromosome Location:
Chr.18: 36560603 - 36658908 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01139030_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ankhd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175375.3 NP_780584.2 18 - 19 3558 115
GenBank mRNA AK017018.1 - 8 - 9 897 115
AK029322.1 - 11 - 12 1973 115
AK042754.1 - 5 - 6 533 115
AK131166.1 - 18 - 19 3421 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000332.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ankhd1

Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800002527.mm.1 NM_175375.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800002527.mm.1 NM_175375.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800002527.mm.1 TC1800000332.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0