This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00849906_g1
See other RPS14 GE Assays ›
Gene Symbol:
Rps14
Gene Name:
ribosomal protein S14
Gene Aliases:
2600014J02Rik, AL023078
Chromosome Location:
Chr.18: 60774596 - 60778546 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00849906_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020600.4 NP_065625.2 5 541 85
GenBank mRNA AK003148.1 - 5 486 85
AK011213.1 - 5 486 85
AK012425.1 - 5 485 85
AK012584.1 - 5 485 85
AK019233.1 - 5 484 85
AK028106.1 - 5 489 85
AK146325.1 - 5 482 85
AK146976.1 - 5 485 85
AK152461.1 - 1 3844 85
AK153401.1 - 1 3844 85
AK168237.1 - 5 485 85
BC042940.1 - 4 465 6487681 85
BC062874.1 - 4 464 5683900 85
BC081449.1 - 4 452 5684375 85

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps14

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
negative regulation of transcription from RNA polymerase II promoter
translation
regulation of translation
erythrocyte differentiation
structural constituent of ribosome
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753488 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753364 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753493 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753490 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753361 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753492 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753489 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753363 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186672 AK168237.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186672 AV490910.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186672 BC042940.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186672 BC062874.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186672 BC081449.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329273 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196510 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174449 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186672 AK168237.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186672 AV490910.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186672 BC042940.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186672 BC062874.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186672 BC081449.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361638 AK168237.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361638 BC042940.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361638 BC062874.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361638 BC081449.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361638 BY149058.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753493 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753476 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753492 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753491 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753499 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753486 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753478 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753498 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753481 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753507 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753473 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753502 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753485 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753487 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753506 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753483 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753490 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753495 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753497 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753480 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753477 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753489 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753472 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31753503 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174449 - -
Whole Mouse Genome G4122A A_51_P361638 AK168237.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P361638 BC042940.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P361638 BC062874.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P361638 BC081449.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P361638 BY149058.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134025 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109987 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174449 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13874151 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13874143 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13874170 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13874149 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13874153 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753361 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753488 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753492 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753490 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753364 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753489 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753493 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31753363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753550 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753539 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753478 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753563 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753499 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753530 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753495 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31753490 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13761587 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753542 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13874159 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13761588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753539 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753455 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13841917 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14136036 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753453 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753487 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753512 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753505 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753502 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753497 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753466 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753546 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753545 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753518 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753527 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753480 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753532 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13985527 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753509 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753482 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753503 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13874149 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753483 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753538 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753452 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753563 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753476 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753486 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753485 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753529 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753515 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14129872 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13668753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753543 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13874143 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753536 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753473 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753499 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753470 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753525 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13874174 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753471 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13742297 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753498 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13798586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753458 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13841915 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13632647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753520 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13614864 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13914559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13633877 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14111377 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31753495 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10456002 - -
Affymetrix Mouse Gene 1.0 ST Array 10456004 - -
Affymetrix Mouse Gene 1.0 ST Array 10456003 - -
Affymetrix Mouse Gene 1.0 ST Array 10498157 - -
Affymetrix Mouse Gene 1.0 ST Array 10498156 - -
Affymetrix HT MG-430 PM Array Plate 1436586_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10456001 - -
Affymetrix Mouse Gene 1.1 ST Array 10498155 - -
Mouse Genome 430 2.0 Array 1436586_PM_x_at - -
Mouse Genome 430 2.0 Array 1437706_PM_x_at - -
Mouse Genome 430 2.0 Array 1437706_x_at - -
Mouse Genome 430 2.0 Array 1436586_x_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20044_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20044_at - -
Affymetrix Mouse Clariom S Array PSR1800004384.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004384.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004384.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004397.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004397.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004397.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004400.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004400.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004400.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004396.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004396.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004396.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004401.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004401.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004401.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004402.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004402.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004402.mm.1 TC1800000616.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004399.mm.1 NM_020600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004399.mm.1 NM_020600.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800004399.mm.1 TC1800000616.mm.1 MTA TC
Mouse Expression Set 430 1436586_x_at - -
Mouse Expression Set 430 1436586_PM_x_at - -
Mouse Expression Set 430 1437706_x_at - -
Mouse Expression Set 430 1437706_PM_x_at - -
Murine Genome U74v2 Set 99657_at - -
Murine Genome U74v2 Set 99657_PM_at - -
Murine Genome U74v2 Set 171270_PM_r_at - -
Murine Genome U74v2 Set 171270_r_at - -
Mouse Genome 430A 2.0 Array 1436586_x_at - -
Mouse Genome 430A 2.0 Array 1437706_PM_x_at - -
Mouse Genome 430A 2.0 Array 1437706_x_at - -
Mouse Genome 430A 2.0 Array 1436586_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1436586_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1436586_x_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 891904 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 430541 - -
Illumina mouse-6 v1.0 expression beadchip 430541 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000430541 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712642 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2712642 - -
mouse-6 v1.1 (Illumina) scl020044.5_19-S - -
Illumina Sentrix mouse-6.v1 bead array 430541 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl020044.5_19-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 430541 - -
MouseRef-8 v2 Expression BeadChip ILMN_2712642 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712642 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020044.5_19-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0